3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)

Lieferant: EHRENSTORFER GMBH

EHERC20012600EA 145 CHF
EHERC20012600
3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)
3,3',4,4',5-Pentachlorbiphenyl (PCB Nr. 126)
Formel: C₁₂H₅Cl₅
MW: 326,44 g/mol
CAS-Nummer: 57465-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR