2,2',3,4,5',6-Hexachlorbiphenyl (PCB Nr. 144)
Lieferant: LGC Standards PROMOCHEM
PROOCIL-PCB-144-CEA
1420
CHF
PROOCIL-PCB-144-C
2,2',3,4,5',6-Hexachlorbiphenyl (PCB Nr. 144)
2,2',3,4,5',6-Hexachlorbiphenyl (PCB Nr. 144)
Formel:
C₁₂H₄Cl₆ MW: 360,88 g/mol |
CAS-Nummer:
68194-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...