Formaldehyd 32% aqueous solution, EM Grade

Gefahr

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

15714-S 15714
ELMI15714-SEA 0 CHF
ELMI15714-S ELMI15714
Formaldehyd 32% aqueous solution, EM Grade
Formaldehyd

Paraformaldehyde - methanol free solution.


  • Microfiltered
  • Stored in pre-scored ampoules


A more efficient and rapid penetrant fixative used in combination with glutaraldehyde and acrolein fixatives.


Easy to break, prescored, 10 ml ampoules sealed under inert gas, or 100 ml bottles.

Formel: CH₂O
MW: 30.03 g/mol
Siedepunkt: >93 °C (>199 °F)
Melting Pt: -92
Dichte: 0.9392 g/cm³ (7.838 lbs/gal.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003274
CAS-Nummer: 50-00-0
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR