6-Chlor-m-toluidin

Lieferant: EHRENSTORFER GMBH

EHERC11429500EA 68.9 CHF
EHERC11429500
6-Chlor-m-toluidin
6-Chlor-m-toluidin
Formel: C₇H₈ClN
MW: 141,6 g/mol
Siedepunkt: 228…230 °C (1013 hPa)
Melting Pt: 30 °C
MDL Number: MFCD00007674
CAS-Nummer: 95-81-8
EINECS: 202-454-7
UN: 2239
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR