Sulfanilamid

Lieferant: Roth Carl

Synonyms: Anilin-4-sulfonsäureamid4-Aminobenzolsulfonsäureamid4-Aminobenzolsulfonamid

ROTH4716.1EA 49.3 CHF
ROTH4716.1
Sulfanilamid
Sulfanilamid
Formel: H₂NC₆H₄SO₂NH₂
MW: 172,21 g/mol
Siedepunkt: 401 °C (1013 hPa)
Melting Pt: 163…167 °C
Dichte: 1,46 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007939
CAS-Nummer: 63-74-1
EINECS: 200-563-4
Merck Index: 12,09094
https://ch.vwr.com/stibo/web/std.lang.all/76/03/2297603.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR