3-Fluorphenylboronsäure

Lieferant: Molekula
Achtung

Synonyms: Dihydroxy(3-fluorphenyl)boran3-Fluorbenzolboronsäure(3-Fluorphenyl)metaborsäure

MOLEM76507241EA 0 CHF
MOLEM76507241
3-Fluorphenylboronsäure
3-Fluorphenylboronsäure
Formel: C₆H₆BFO₂
MW: 139,92 g/mol
Melting Pt: 216…218 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00236042
CAS-Nummer: 768-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR