N-Hydroxysuccinimid

Lieferant: Molekula

Synonyms: 1-Hydroxy-1,4-butandisäureimid1-Hydroxy-2,5-pyrrolidindion1-HydroxybernsteinsäureimidNHSHOSuN-Hydroxy-1,4-butandisäureimidN-Hydroxybernsteinsäureimid1-Hydroxyazacyclopentan-2,5-dion

MOLEM24112630EA 0 CHF
MOLEM24112630
N-Hydroxysuccinimid
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
MW: 115,09 g/mol
Siedepunkt: 363 °C (1013 hPa)
Melting Pt: 93…95 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR