L(-)-Glutathion (oxidierte Form)
Lieferant: EUROCLONE
Synonyms:
L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)
EMR261001
EMR261005
EMR261025
EURLEMR261001EA
0
CHF
EURLEMR261001
EURLEMR261005
EURLEMR261025
L(-)-Glutathion (oxidierte Form)
L(-)-Glutathion (oxidierte Form)
Formel:
C₂₀H₃₂N₆O₁₂S₂ MW: 612.64 g/mol Melting Pt: 185 Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00063106 CAS-Nummer: 27025-41-8 EINECS: 248-170-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...