L(-)-Glutathion (reduzierte Form)
Lieferant: EUROCLONE
Synonyms:
L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
EMR262005
EMR262025
EMR262100
EURLEMR262005EA
0
CHF
EURLEMR262005
EURLEMR262025
EURLEMR262100
L(-)-Glutathion (reduzierte Form)
L(-)-Glutathion (reduzierte Form)
Formel:
C₁₀H₁₇N₃O₆S Melting Pt: 185…195 °C Dichte: 1.475 g/cm³ (20 °C) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00065939 CAS-Nummer: 70-18-8 EINECS: 200-725-4 Merck Index: 13,04488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...