1-Dodecansulfonsäure Natriumsalz 99+%

Lieferant: Thermo Scientific

Synonyms: NatriumlaurylsulfonatNatriumdodecylsulfonat

ACRO147900010EA 32.5 CHF
ACRO147900010
1-Dodecansulfonsäure Natriumsalz 99+%
1-Dodecansulfonsäure Natriumsalz
Formel: C₁₂H₂₅NaO₃S
MW: 272,38 g/mol
Melting Pt: >300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007527
CAS-Nummer: 2386-53-0
EINECS: 219-200-6

Order Now

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR