Dithiothreitol (DTT, Cleland's Reagenz)

Lieferant: EUROCLONE
Achtung

EMR033005 EMR033010 EMR033025
EURLEMR033005EA 0 CHF
EURLEMR033005 EURLEMR033010 EURLEMR033025
Dithiothreitol (DTT, Cleland's Reagenz)
Dithiothreitol (DTT, Cleland's Reagenz)
Formel: C₄H₁₀O₂S₂
Siedepunkt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash-Pt: >110 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00004877
CAS-Nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR