2-Propanol

Lieferant: EUROCLONE
Gefahr

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

EMR075001 EMR075025
EURLEMR075001EA 0 CHF
EURLEMR075001 EURLEMR075025
2-Propanol
2-Propanol
Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Siedepunkt: 82
Melting Pt: –89 °C
Dichte: 0.786 g/cm³ (20 °C)
Flash-Pt: 12 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR