Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)

Lieferant: GERBU BIOCHEMICALS
Achtung

1018.0250 1018.1000 1018.2500 1018.5000
GRBU1018.0250EA 0 CHF
GRBU1018.0250 GRBU1018.1000 GRBU1018.2500 GRBU1018.5000
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethan (TRIS, Trometamol)
Formel: NH₂C(CH₂OH)₃
Siedepunkt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Dichte: 1.328 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004679
CAS-Nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR