N,N,N',N'-Tetramethylethylendiamin (TEMED)

Lieferant: NATIONAL DIAGNOSTICS
Gefahr

Synonyms: 1,2-Bis(dimethylamino)ethan

NADIEC-503-25MLEA 0 CHF
NADIEC-503-25ML
N,N,N',N'-Tetramethylethylendiamin (TEMED)
N,N,N',N'-Tetramethylethylendiamin (TEMED)
Formel: C₆H₁₆N₂
MW: 116,21 g/mol
Siedepunkt: 120 °C (1013 hPa)
Melting Pt: –64 °C
Dichte: 0,7765 g/cm³ (20 °C)
Flash-Pt: 17 °C (geschlossener Tiegel)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008335
CAS-Nummer: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://ch.vwr.com/stibo/web/std.lang.all/99/66/2299966.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR