N,N,N',N'-Tetramethylethylendiamin (TEMED)

Lieferant: EUROCLONE
Gefahr

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine

EMR228100 EMR228250 EMR228500
EURLEMR228100EA 0 CHF
EURLEMR228100 EURLEMR228250 EURLEMR228500
N,N,N',N'-Tetramethylethylendiamin (TEMED)
N,N,N',N'-Tetramethylethylendiamin (TEMED)
Formel: C₆H₁₆N₂
MW: 116,21 g/mol
Siedepunkt: 120 °C (1013 hPa)
Melting Pt: –64 °C
Dichte: 0.7765 g/cm³ (20 °C)
Flash-Pt: 17 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008335
CAS-Nummer: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR