Iodacetamid
Lieferant: G Bioscienes
|
Achtung
|
Synonyms:
Iodessigsäureamid
786-078EA
565
CHF
786-078
Iodacetamid
Iodacetamid
Formel:
ICH₂CONH₂ MW: 184,96 g/mol Siedepunkt: 266 °C (1013 hPa) Melting Pt: 91…94 °C Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00008028 CAS-Nummer: 144-48-9 EINECS: 205-630-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...