Glycin (H-Gly-OH)
Lieferant: GERBU BIOCHEMICALS GMBH
Synonyms:
H-Gly-OH
, Glycocoll
, Aminoessigsäure
GRBU1023.0250EA
0
CHF
GRBU1023.0250
GRBU1023.1000
GRBU1023.5000
Glycin (H-Gly-OH)
Glycin (H-Gly-OH)
Formel:
NH₂CH₂COOH MW: 75,07 g/mol Siedepunkt: 181 °C (1013 hPa) Melting Pt: 83 °C Dichte: 1,64 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00008131 CAS-Nummer: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...