Glycin (H-Gly-OH)

Lieferant: Alpha Diagnostics International

Synonyms: H-Gly-OHGlycocollAminoessigsäure

ALPDGLYC51-SEA 0 CHF
ALPDGLYC51-S
Glycin (H-Gly-OH)
Glycin (H-Gly-OH)
Formel: NH₂CH₂COOH
MW: 75,07 g/mol
Siedepunkt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Dichte: 1,64 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008131
CAS-Nummer: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://ch.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR