4-Aminobuttersäure
Lieferant: Chemicon
Synonyms:
GABA, γ-Aminobutyric acid
AB5592-200UL
CHMCAB5592-200ULEA
2340
CHF
CHMCAB5592-200UL
4-Aminobuttersäure
4-Aminobuttersäure
Formel:
C₄H₉NO₂ Siedepunkt: 248 °C (1013 hPa) Melting Pt: 202 °C Dichte: 1.21 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00008226 CAS-Nummer: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...