N-Lauroylsarkosin Natriumsalz, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Gefahr
|
Synonyms:
Sarkosyl
, Natrium-N-lauroylsarkosinat
, N-Dodecanoyl-N-methylglycin Natriumsalz
, Lauroylsarkosinat
428010-5EA
642
CHF
428010-5
N-Lauroylsarkosin Natriumsalz, Millipore®
N-Lauroylsarkosin Natriumsalz
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa MW: 293,38 g/mol Melting Pt: 45…47 °C Flash-Pt: >150 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00042728 CAS-Nummer: 137-16-6 EINECS: 205-281-5 UN: 2811 ADR: 6.1,II Merck Index: 12,04379 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...