Diethylsuccinat

Lieferant: Molekula

MOLE13652039-1KGEA 0 CHF
MOLE13652039-1KG
Diethylsuccinat
Diethylsuccinat
Formel: C₈H₁₄O₄
MW: 174,2 g/mol
Siedepunkt: 217 °C (1013 hPa)
Melting Pt: –19 °C
Dichte: 1,04 g/cm³ (20 °C)
Flash-Pt: 80 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009208
CAS-Nummer: 123-25-1
EINECS: 204-612-0
Merck Index: 12,09038

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR