N,N-Dimethylformamid

Lieferant: EUROCLONE
Gefahr

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

EMR177001 EMR177025 EMR177500
EURLEMR177001EA 0 CHF
EURLEMR177001 EURLEMR177025 EURLEMR177500
N,N-Dimethylformamid
N,N-Dimethylformamid
Formel: HCON(CH₃)₂
Siedepunkt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Dichte: 0.949 g/cm³ (20 °C)
Flash-Pt: 57.5 °C (closed cup)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003284
CAS-Nummer: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR