Fluorescein-12-UTP
Lieferant: Enzo Life Sciences
ALEXENZ-42834EA 494 CHF
ALEXENZ-42834
Fluorescein-12-UTP
Proteinkonjugate und Markierungen
Fluorescein-12-UTP is ideal for in vitro transcription reactions catalysed by T3, T7, or SP6 RNA polymerases and reverse transcriptase to produce fluorophore-labeled RNA transcripts. RNA-based probes can be used to identify complementary sequences by in situ hybridisation to fixed cells and tissues by direct or indirect fluorescence detection or by membrane hybridisation procedures.