Natriumpyruvat

Lieferant: EUROCLONE

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

EMR213001 EMR213100 EMR213500
EURLEMR213001EA 0 CHF
EURLEMR213001 EURLEMR213100 EURLEMR213500
Natriumpyruvat
Natriumpyruvat
Formel: CH₃COCOONa
MW: 110.04 g/mol
Melting Pt: 220…230 °C
Dichte: 1.718 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002586
CAS-Nummer: 113-24-6
EINECS: 204-024-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR