1,10-Phenanthrolin Monohydrat ACS
Lieferant: VWR Chemicals
|
Gefahr
|
0516-10GEA
175
CHF
0516-10G
1,10-Phenanthrolin Monohydrat ACS
1,10-Phenanthrolin Monohydrat
A metalloprotease inhibitor useful for chelating iron and other divalent metals.
Formel:
C₁₂H₈N₂·1H₂O MW: 198,22 g/mol Melting Pt: 93…94 °C Dichte: 1,259 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00149973 CAS-Nummer: 5144-89-8 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III Merck Index: 13,07291 |
Spezifikation Testergebnisse
Suitability as Redox Indicator | Pass |
Suitability for Determining Iron | pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...