D(+)-Biotin

Lieferant: EUROCLONE

EMR257001 EMR257250 EMR257500
EURLEMR257001EA 0 CHF
EURLEMR257001 EURLEMR257250 EURLEMR257500
D(+)-Biotin
D(+)-Biotin
Formel: C₁₀H₁₆N₂O₃S
Melting Pt: 232…233 °C
Dichte: 1,41 g/cm³ (20 °C)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00005541
CAS-Nummer: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR