D(+)-Biotin
Lieferant: EUROCLONE
Synonyms:
(3aS,4S,6aR)-Biotin
, 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H- thieno[3,4-d]imidazol-4-yl]pentansäure
, Vitamin H
, Vitamin B8
, Coenzym R
, Vitamin B7
EURLEMR115001EA
0
CHF
EURLEMR115001
EURLEMR115250
EURLEMR115500
D(+)-Biotin
D(+)-Biotin
Formel:
C₁₀H₁₆N₂O₃S MW: 244,31 g/mol Melting Pt: 232…233 °C Dichte: 1,41 g/cm³ (20 °C) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00005541 CAS-Nummer: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...