L(-)-Tyrosin

Lieferant: Molekula

Synonyms: (S)-(-)-2-Amino-3-(4-hydroxyphenyl)propionsäureL(-)-p-HydroxyphenylalaninYTyr(S)-(-)-p-HydroxyphenylalaninH-Tyr-OH

MOLE12097155-1KGEA 0 CHF
MOLE12097155-1KG
L(-)-Tyrosin
L(-)-Tyrosin
Formel: C₉H₁₁NO₃
MW: 181,19 g/mol
Siedepunkt: 351 °C (1013 hPa)
Melting Pt: 285 °C
Dichte: 1,41 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002606
CAS-Nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 12,09970

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR