D(+)-Tyrosin

Lieferant: Molekula

Synonyms: D(+)-p-Hydroxyphenylalanin(R)-(+)-2-Amino-3-(4-hydroxyphenyl)propionsäureH-D-Tyr-OH(R)-(+)-p-Hydroxyphenylalanin

MOLEM24028905EA 0 CHF
MOLEM24028905
D(+)-Tyrosin
D(+)-Tyrosin
Formel: C₉H₁₁NO₃
MW: 181,19 g/mol
Siedepunkt: 351 °C (1013 hPa)
Melting Pt: 285 °C
MDL Number: MFCD00063073
CAS-Nummer: 556-02-5
EINECS: 209-112-6
Merck Index: 12,09970

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR