L(+)-Leucin, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyms:
(S)-(+)-Leucin
, (S)-(+)-2-Amino-4-methylpentansäure
, L-Leucin
, H-Leu-OH
, (S)-(+)-α-Amino-γ-methylvaleriansäure
4330-100EA
212
CHF
4330-100
L(+)-Leucin, Millipore®
L(+)-Leucin
Formel:
C₆H₁₃NO₂ MW: 131,17 g/mol Melting Pt: 300 °C Dichte: 1,165 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00002617 CAS-Nummer: 61-90-5 EINECS: 200-522-0 Merck Index: 14,05451 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...