Eisen(II)sulfat Heptahydrat
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
Iron vitriol, Ferrous sulphate heptahydrate
EMR278001
EMR278005
EMR278500
EURLEMR278001EA
0
CHF
EURLEMR278001
EURLEMR278005
EURLEMR278500
Eisen(II)sulfat Heptahydrat
Eisen(II)sulfat Heptahydrat
Formel:
FeSO₄·7H₂O Melting Pt: 64 °C Dichte: 1.9 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00149719 CAS-Nummer: 7782-63-0 EINECS: 231-753-5 Merck Index: 13,04091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...