Mangan(II)sulfat Monohydrat
Lieferant: EUROCLONE
|
Achtung
|
Synonyms:
Manganese sulphate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate
EMR279001
EMR279250
EMR279500
EURLEMR279001EA
0
CHF
EURLEMR279001
EURLEMR279250
EURLEMR279500
Mangan(II)sulfat Monohydrat
Mangan(II)sulfat Monohydrat
Formel:
MnSO₄·1H₂O MW: 169.02 g/mol Melting Pt: 700 Dichte: 2.95 g/cm³ (25 °C) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00149159 CAS-Nummer: 10034-96-5 EINECS: 232-089-9 UN: 3077 ADR: 9 III Merck Index: 13,05763 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...