Control RNA
Lieferant: Enzo Life Sciences
ALEXENZ-42406-CREA 0 CHF
ALEXENZ-42406-CR
Control RNA
Nukleinsäure-Reagenzien Modifikation und Markierung von Nukleinsäure
Control RNA is prepared from homogenized HeLa-S3 cells.
  • Ideal for use as a positive control with single round RNA amplification and labeling system
  • Also useful in RT-PCR, Northern analysis, ribonuclease protection assays, cDNA synthesis and in vitro translation
Concentration: 100 ng/µl