EDTA Dinatriumsalz Dihydrat

Lieferant: GERBU BIOCHEMICALS

1034.0250 1034.1000
GRBU1034.0250EA 0 CHF
GRBU1034.0250 GRBU1034.1000
EDTA Dinatriumsalz Dihydrat
EDTA Dinatriumsalz Dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00150037
CAS-Nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR