Guanidiniumthiocyanat

Lieferant: EUROCLONE
Achtung

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

EMR017001 EMR017100 EMR017500
EURLEMR017001EA 0 CHF
EURLEMR017001 EURLEMR017100 EURLEMR017500
Guanidiniumthiocyanat
Guanidiniumthiocyanat
Formel: CH₅N₃·CHNS
Melting Pt: 118 °C
Dichte: 1.29 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013027
CAS-Nummer: 593-84-0
EINECS: 209-812-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR