Guanidiniumchlorid

Lieferant: EUROCLONE
Achtung

EMR149001 EMR149250 EMR149500
EURLEMR149001EA 0 CHF
EURLEMR149001 EURLEMR149250 EURLEMR149500
Guanidiniumchlorid
Guanidiniumchlorid
Formel: NH₂C(=NH)NH₂·HCl
Melting Pt: 185 °C
Dichte: 1.354 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013026
CAS-Nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR