TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)

Lieferant: GERBU BIOCHEMICALS

1028.0100 1028.0250 1028.1000 1028.5000
GRBU1028.0100EA 0 CHF
GRBU1028.0100 GRBU1028.0250 GRBU1028.1000 GRBU1028.5000
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
TRIS HCl (Tris(hydroxymethyl)aminomethan Hydrochlorid)
Formel: NH₂C(CH₂OH)₃·HCl
Melting Pt: 150…152 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012590
CAS-Nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR