(±)-3-Chinuclidinol

Lieferant: LGC Standards PROMOCHEM

Synonyms: (±)-Chinuclidin-3-ol(±)-3-Hydroxychinuclidin(±)-3-Hydroxy-1-azabicyclo[2.2.2]octan

PROOMM0825.02EA 2360 CHF
PROOMM0825.02
(±)-3-Chinuclidinol
(±)-3-Chinuclidinol
Formel: C₇H₁₃NO
MW: 127,19 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD00151326
CAS-Nummer: 1619-34-7
EINECS: 216-578-4
UN: 3263
ADR: 8,III
Merck Index: 12,08266

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR