(±)-3-Chlor-1,2-propandiol

Lieferant: LGC Standards PROMOCHEM

Synonyms: (±)-3-Chlor-1,2-dihydroxypropan

PROOMM0027.22EA 574 CHF
PROOMM0027.22
(±)-3-Chlor-1,2-propandiol
(±)-3-Chlor-1,2-propandiol
Formel: C₃H₇ClO₂
MW: 110,54 g/mol
Siedepunkt: 213 °C (1013 hPa)
Melting Pt: –40 °C
Dichte: 1,32 g/cm³ (20 °C)
Flash-Pt: 135 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004712
CAS-Nummer: 96-24-2
EINECS: 202-492-4
UN: 2689
ADR: 6.1,III
Merck Index: 13,02164

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR