(±)-Linalool

Lieferant: Roth Carl
Achtung

Synonyms: (±)-3,7-Dimethyl-3-hydroxy-1,6-octadien(±)-3,7-Dimethyl-1,6-octadien-3-ol

ROTH3992.1EA 34.45 CHF
ROTH3992.1
(±)-Linalool
(±)-Linalool
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Siedepunkt: 199 °C (1013 hPa)
Dichte: 0,87 g/cm³ (20 °C)
Flash-Pt: 75 °C
Lager Temperatur: Raumtemperatur
CAS-Nummer: 78-70-6
EINECS: 201-134-4
Merck Index: 13,05517

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR