Tween® 20 (Polysorbat)
Lieferant: EUROCLONE
Synonyms:
Polyoxyethylene sorbitan monolaurate
EMR243001
EMR243005
EMR243025
EMR243500
EURLEMR243001EA
0
CHF
EURLEMR243001
EURLEMR243005
EURLEMR243025
EURLEMR243500
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)
Formel:
C₅₈H₁₁₄O₂₆ MW: 1227.53 g/mol Dichte: 1.1 g/cm³ (20 °C) Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...