TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
Lieferant: EUROCLONE
|
Gefahr
|
Synonyms:
Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
EMR237001
EMR237005
EMR237025
EMR237500
EURLEMR237001EA
0
CHF
EURLEMR237001
EURLEMR237005
EURLEMR237025
EURLEMR237500
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
Siedepunkt:
∼200 °C (1013 hPa) Melting Pt: 6 °C Dichte: 1.06 g/cm³ (20 °C) Flash-Pt: 251 °C (closed cup) Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00132505 CAS-Nummer: 9002-93-1 UN: 3082 ADR: 9,III Merck Index: 13,06793 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...