Streptomycinsulfat

Lieferant: EUROCLONE
Achtung

EMR218025 EMR218100 EMR218250
EURLEMR218025EA 0 CHF
EURLEMR218025 EURLEMR218100 EURLEMR218250
Streptomycinsulfat
Streptomycin sulphate
Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Melting Pt: 250 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00037023
CAS-Nummer: 3810-74-0
EINECS: 223-286-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,08983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR