TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in Wasser

Lieferant: EUROCLONE
Gefahr

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

EMR238025 EMR238100
EURLEMR238025EA 0 CHF
EURLEMR238025 EURLEMR238100
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether) 10% in Wasser
TRITON™ X-100 (Polyethylenglycol-tert-octylphenylether)
Siedepunkt: ∼100 °C (1013 hPa)
Melting Pt: 6
Dichte: ∼1.01 g/cm³ (20 °C)
Flash-Pt: 251
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00132505
CAS-Nummer: 9002-93-1
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR