D(+)-Saccharose

Lieferant: EUROCLONE

EMR119001 EMR119500
EURLEMR119001EA 0 CHF
EURLEMR119001 EURLEMR119500
D(+)-Saccharose
D(+)-Saccharose
Formel: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Dichte: 1.29 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR