D(+)-Glucose Monohydrat

Lieferant: EUROCLONE

Synonyms: Dextrose MonohydratTraubenzucker Monohydrat

EURLEMR295001EA 0 CHF
EURLEMR295001
D(+)-Glucose Monohydrat
D(+)-Glucose Monohydrat
Formel: C₆H₁₂O₆·H₂O
MW: 198,17 g/mol
Melting Pt: 83 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149450
CAS-Nummer: 14431-43-7
EINECS: 200-075-1
https://ch.vwr.com/stibo/web/std.lang.all/21/23/2302123.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR