D(+)-Glucose Monohydrat
Lieferant: EUROCLONE
Synonyms:
(2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
EMR116001
EMR116500
EURLEMR116001EA
0
CHF
EURLEMR116001
EURLEMR116500
D(+)-Glucose Monohydrat
D(+)-Glucose Monohydrat
Formel:
C₆H₁₂O₆·H₂O MW: 198.17 g/mol Melting Pt: 83 Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00149450 CAS-Nummer: 14431-43-7 EINECS: 200-075-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...