D(+)-Trehalose Dihydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyms:
Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
A19434.06
A19434.14
A19434.22
A19434.06EA
60.1
CHF
A19434.06
A19434.14
A19434.22
D(+)-Trehalose Dihydrat ≥99%
D(+)-Trehalose Dihydrat
Purity: 99%
CAS No.: 6138-23-4
Molecular Formula: C12H22O11.2H2O
Formula Weight: 378.33 (342.29anhy)
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 97-99°
Beilstein Reference: 5322018
Merck Reference: 14,9580
Optical Rotation: +179° (c=2 in water)
MDL No.: MFCD00071594
Formel:
C₁₂H₂₂O₁₁·2H₂O MW: 378.33 g/mol Melting Pt: 97…99 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00071594 CAS-Nummer: 6138-23-4 EINECS: 202-739-6 Merck Index: 14,9580 |