D(+)-Maltose Monohydrat

Lieferant: EUROCLONE

Synonyms: Malzzucker

EURLEMR299500EA 0 CHF
EURLEMR299500
D(+)-Maltose Monohydrat
D(+)-Maltose Monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
MW: 360,32 g/mol
Melting Pt: 160…165 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149343
CAS-Nummer: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://ch.vwr.com/stibo/web/std.lang.all/21/24/2302124.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR