Cisplatin, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyms: cis-Diammindichloridoplatin(II)Platinol

232120-50EA 246 CHF
232120-50
Cisplatin, Millipore®
cis-Platin
Formel: H₆Cl₂N₂Pt
MW: 300,05 g/mol
Dichte: 3,738 g/cm³ (20 °C)
MDL Number: MFCD00011623
CAS-Nummer: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR